Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL18R1 Antibody (MM0380-8H22), Allophycocyanin, Novus Biologicals™
SDP

Catalog No. NB344393 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB344393 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB344393 Supplier Novus Biologicals Supplier No. NBP211679APC
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

IL18R1 Monoclonal antibody specifically detects IL18R1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Neutralization

Specifications

Antigen IL18R1
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Neutralization
Classification Monoclonal
Clone MM0380-8H22
Conjugate APC
Dilution Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Block/Neutralize
Formulation PBS
Gene Alias CD218 antigen-like family member A, CD218a, CD218a antigen, CDw218a, cytokine receptor, IL-18R1, IL-18R-1, IL1R-rp, IL1RRPIL1 receptor-related protein, IL-1RrpIL18RA, interleukin 18 receptor 1, interleukin-18 receptor 1
Host Species Mouse
Immunogen Human recombinant IL18R1 (extracellular domain)
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline CD Markers
Primary or Secondary Primary
Gene ID (Entrez) 8809
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.