Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL18R1 Antibody (MM0380-8H22) - Azide and BSA Free, Novus Biologicals™
SDP

Catalog No. NB018850 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB018850 0.025 mg
NBP211679 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB018850 Supplier Novus Biologicals Supplier No. NBP2116790.025MG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

IL18R1 Monoclonal specifically detects IL18R1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Block/Neutralize.

Specifications

Antigen IL18R1
Applications Blocking Assay, Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone MM0380-8H22
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500, Blocking/Neutralizing 50-200 molar excess
Formulation A 0.2 um filtered solution in PBS with No Preservative
Gene Accession No. Q13478
Gene Alias CD218 antigen-like family member A, CD218a, CD218a antigen, CDw218a, cytokine receptor, IL-18R1, IL-18R-1, IL1R-rp, IL1RRPIL1 receptor-related protein, IL-1RrpIL18RA, interleukin 18 receptor 1, interleukin-18 receptor 1
Gene Symbols IL18R1
Host Species Mouse
Immunogen Produced from a hybridoma (mouse myeloma fused with spleen cells from a mouse immunized with human recombinant protein of IL-18 Ra extracellular domain
Purification Method Protein G purified
Quantity 0.025 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8809
Reconstitution Reconstitute with 0.2 ml sterilized PBS to a final concentration of 0.5 mg/ml.
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.