Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IL18R1 Antibody (MM0380-8H22) - Azide and BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
$206.50 - $560.50
Specifications
Antigen | IL18R1 |
---|---|
Clone | MM0380-8H22 |
Concentration | LYOPH |
Applications | Blocking Assay, Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin) |
Classification | Monoclonal |
Description
IL18R1 Monoclonal specifically detects IL18R1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Block/Neutralize.Specifications
IL18R1 | |
LYOPH | |
Monoclonal | |
Purified | |
RUO | |
Q13478 | |
8809 | |
IgG1 | |
Protein G purified |
MM0380-8H22 | |
Blocking Assay, Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Mouse | |
Human | |
CD218 antigen-like family member A, CD218a, CD218a antigen, CDw218a, cytokine receptor, IL-18R1, IL-18R-1, IL1R-rp, IL1RRPIL1 receptor-related protein, IL-1RrpIL18RA, interleukin 18 receptor 1, interleukin-18 receptor 1 | |
IL18R1 | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title