Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL18R1 Antibody (MM0380-8H22), DyLight 405, Novus Biologicals™
SDP

Catalog No. NBP211679V Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP211679V 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP211679V Supplier Novus Biologicals Supplier No. NBP211679V
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

IL18R1 Monoclonal specifically detects IL18R1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen IL18R1
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone MM0380-8H22
Conjugate DyLight 405
Dilution Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias CD218 antigen-like family member A, CD218a, CD218a antigen, CDw218a, cytokine receptor, IL-18R1, IL-18R-1, IL1R-rp, IL1RRPIL1 receptor-related protein, IL-1RrpIL18RA, interleukin 18 receptor 1, interleukin-18 receptor 1
Gene Symbols IL18R1
Host Species Mouse
Immunogen Human recombinant IL18R1 (extracellular domain)
Purification Method Protein G purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 8809
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.