Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ IL18R1 Recombinant Rabbit Monoclonal Antibody (BLR172J)
Rabbit Recombinant Monoclonal Antibody
Supplier: Invitrogen™ MA544529
This item is not returnable.
View return policy
Description
IL18R1 Recombinant Monoclonal Antibody for Western Blot, Flow
IL18R1 is a cytokine receptor that belongs to the interleukin 1 receptor family. This receptor specifically binds interleukin 18 (IL18), and is essential for IL18 mediated signal transduction. IFN-alpha and IL12 are reported to induce the expression of this receptor in NK and T cells. The gene along with four other members of the interleukin 1 receptor family, including IL1R2, IL1R1, ILRL2 (IL-1Rrp2), and IL1RL1 (T1/ST2), form a gene cluster on chromosome 2q.
Specifications
IL18R1 | |
Recombinant Monoclonal | |
100 μg/mL | |
BBS with 0.1% BSA and 0.09% sodium azide; pH 7.8-8.6 | |
Q13478 | |
Il18r1 | |
Recombinant protein representing the extracellular domain of Human Interleukin-18 receptor 1 (residues 25-329). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Flow Cytometry, Western Blot | |
BLR172J | |
Unconjugated | |
Il18r1 | |
CD218 antigen-like family member A; CD218a; CDw218a; cytokine receptor; IL1 receptor-related protein; Il18r1; IL-18R1; IL-18R-1; IL18RA; Il18ralpha; IL-18R-alpha; IL18Ralpha2; IL1RRP; IL-1Rrp; IL1R-rp; interleukin 1 receptor related protein; interleukin 18 receptor 1; Interleukin-18; interleukin-18 receptor 1; Interleukin-18 receptor alpha | |
Rabbit | |
Purified | |
RUO | |
8809 | |
4°C | |
Liquid |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction