Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ IL18R1 Recombinant Rabbit Monoclonal Antibody (BLR172J)
Rabbit Recombinant Monoclonal Antibody
$173.10 - $436.00
Specifications
Antigen | IL18R1 |
---|---|
Clone | BLR172J |
Concentration | 100 μg/mL |
Content And Storage | 4°C |
Applications | Flow Cytometry, Western Blot |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
PIMA544529
|
Invitrogen™
MA544529 |
100 μL |
Each for $436.00
|
|
|||||
PIMA544530
|
Invitrogen™
MA544530 |
20 μL |
Each for $173.10
|
|
|||||
Description
IL18R1 Recombinant Monoclonal Antibody for Western Blot, Flow
IL18R1 is a cytokine receptor that belongs to the interleukin 1 receptor family. This receptor specifically binds interleukin 18 (IL18), and is essential for IL18 mediated signal transduction. IFN-alpha and IL12 are reported to induce the expression of this receptor in NK and T cells. The gene along with four other members of the interleukin 1 receptor family, including IL1R2, IL1R1, ILRL2 (IL-1Rrp2), and IL1RL1 (T1/ST2), form a gene cluster on chromosome 2q.Specifications
IL18R1 | |
100 μg/mL | |
Flow Cytometry, Western Blot | |
Unconjugated | |
Rabbit | |
RUO | |
BBS with 0.1% BSA and 0.09% sodium azide; pH 7.8-8.6 | |
Q13478 | |
8809 | |
Recombinant protein representing the extracellular domain of Human Interleukin-18 receptor 1 (residues 25-329). | |
Antibody |
BLR172J | |
4°C | |
Recombinant Monoclonal | |
Liquid | |
IgG | |
Human | |
Il18r1 | |
CD218 antigen-like family member A; CD218a; CDw218a; cytokine receptor; IL1 receptor-related protein; Il18r1; IL-18R1; IL-18R-1; IL18RA; Il18ralpha; IL-18R-alpha; IL18Ralpha2; IL1RRP; IL-1Rrp; IL1R-rp; interleukin 1 receptor related protein; interleukin 18 receptor 1; Interleukin-18; interleukin-18 receptor 1; Interleukin-18 receptor alpha | |
Il18r1 | |
Primary | |
Purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title