Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL1RAPL1 Antibody (B-R58), Alexa Fluor™ 532, Novus Biologicals™
SDP

Catalog No. NB382891 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB382891 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB382891 Supplier Novus Biologicals Supplier No. NBP318111AF532
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

IL1RAPL1 Monoclonal antibody specifically detects IL1RAPL1 in Human samples. It is validated for Flow Cytometry, ELISA

Specifications

Antigen IL1RAPL1
Applications Flow Cytometry, ELISA
Classification Monoclonal
Clone B-R58
Conjugate Alexa Fluor 532
Formulation 50mM Sodium Borate
Gene Alias IL1R8IL1RAPL, IL-1-RAPL-1, IL-1RAPL-1, IL1RAPL-1, interleukin 1 receptor accessory protein-like 1, interleukin 1 receptor-8, interleukin-1 receptor accessory protein-like 1, mental retardation, X-linked 10, MRX10, MRX21, MRX34, Oligophrenin-4, OPHN4mental retardation, X-linked 21, Three immunoglobulin domain-containing IL-1 receptor-related 2, TIGIRR-2mental retardation, X-linked 34, X-linked interleukin-1 receptor accessory protein-like 1
Host Species Mouse
Immunogen Recombinant human IL1RAPL1. Hybridoma: Myeloma X63/AG.8653 x Balb/c spleen cells
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11141
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.