Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL1RAPL1 Antibody (MM0353-3R16) - Azide and BSA Free, Novus Biologicals™
SDP

Catalog No. NBP211648 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP211648 0.1 mg
NB018838 0.025 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP211648 Supplier Novus Biologicals Supplier No. NBP211648
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

IL1RAPL1 Monoclonal specifically detects IL1RAPL1 in Human samples. It is validated for Western Blot.

Specifications

Antigen IL1RAPL1
Applications Western Blot
Classification Monoclonal
Clone MM0353-3R16
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1:500-1:2000
Formulation Lyophilized from a 0.2 μm filtered solution in PBS. 0.025 mg size is provided in liquid form, PBS with No Preservative
Gene Accession No. Q9NZN1
Gene Alias IL1R8IL1RAPL, IL-1-RAPL-1, IL-1RAPL-1, IL1RAPL-1, interleukin 1 receptor accessory protein-like 1, interleukin 1 receptor-8, interleukin-1 receptor accessory protein-like 1, mental retardation, X-linked 10, MRX10, MRX21, MRX34, Oligophrenin-4, OPHN4mental retardation, X-linked 21, Three immunoglobulin domain-containing IL-1 receptor-related 2, TIGIRR-2mental retardation, X-linked 34, X-linked interleukin-1 receptor accessory protein-like 1
Gene Symbols IL1RAPL1
Host Species Mouse
Immunogen Human recombinant IL1RAPL1 (extracellular domain)
Purification Method Protein G purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11141
Reconstitution Reconstitute with sterilized PBS to a final concentration of 0.5 mg/ml.
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.