Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL6R Antibody (B-N12), DyLight 650, Novus Biologicals™
SDP

Catalog No. NB388325 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB388325 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB388325 Supplier Novus Biologicals Supplier No. NBP314598C
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

IL6R Monoclonal antibody specifically detects IL6R in Human samples. It is validated for Flow Cytometry, ELISA

Specifications

Antigen IL6R
Applications Flow Cytometry, ELISA
Classification Monoclonal
Clone B-N12
Conjugate DyLight 650
Formulation 50mM Sodium Borate
Gene Alias CD126, CD126 antigen, gp80, IL-6 receptor subunit alpha, IL-6R 1, IL-6R subunit alpha, IL-6R-1, IL6RA, IL-6RA, IL-6R-alpha, interleukin 6 receptor, interleukin 6 receptor alpha subunit, interleukin-6 receptor subunit alpha, Membrane glycoprotein 80, MGC104991
Host Species Mouse
Immunogen gp80 transfected CHO cell line
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Cytokine Research, Immunology, Innate Immunity
Primary or Secondary Primary
Gene ID (Entrez) 3570
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.