Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL6R Antibody (MM0409-6M30), Alexa Fluor™ 647, Novus Biologicals™
SDP

Catalog No. NB211722647 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB211722647 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB211722647 Supplier Novus Biologicals Supplier No. NBP211722AF647

Mouse Monoclonal Antibody

IL6R Monoclonal specifically detects IL6R in Human samples. It is validated for Western Blot.

Specifications

Antigen IL6R
Applications Western Blot
Classification Monoclonal
Clone MM0409-6M30
Conjugate Alexa Fluor 647
Dilution Western Blot
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias CD126, CD126 antigen, gp80, IL-6 receptor subunit alpha, IL-6R 1, IL-6R subunit alpha, IL-6R-1, IL6RA, IL-6RA, IL-6R-alpha, interleukin 6 receptor, interleukin 6 receptor alpha subunit, interleukin-6 receptor subunit alpha, Membrane glycoprotein 80, MGC104991
Gene Symbols IL6R
Host Species Mouse
Immunogen Human recombinant IL6R (extracellular domain)
Purification Method Protein G purified
Quantity 0.1 mL
Research Discipline Cancer, Cytokine Research, Immunology, Innate Immunity
Primary or Secondary Primary
Gene ID (Entrez) 3570
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.