Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL6R Antibody (rhPM-1 (Tocilizumab)), Alexa Fluor™ 350, Novus Biologicals™
SDP

Catalog No. NB151086 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB151086 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB151086 Supplier Novus Biologicals Supplier No. NBP275192AF350
Add to Cart
Add to Cart

Human Monoclonal Antibody

IL6R Monoclonal specifically detects IL6R in Human samples. It is validated for ELISA, Immunofluorescence.

Specifications

Antigen IL6R
Applications ELISA, Immunofluorescence
Classification Monoclonal
Clone rhPM-1 (Tocilizumab)
Conjugate Alexa Fluor 350
Dilution ELISA, Immunofluorescence
Formulation 50mM Sodium Borate
Gene Alias CD126, CD126 antigen, gp80, IL-6 receptor subunit alpha, IL-6R 1, IL-6R subunit alpha, IL-6R-1, IL6RA, IL-6RA, IL-6R-alpha, interleukin 6 receptor, interleukin 6 receptor alpha subunit, interleukin-6 receptor subunit alpha, Membrane glycoprotein 80, MGC104991
Host Species Human
Immunogen This antibody was prepared by CDR grafting of mouse AUK 12-20 antibody onto a human framework regions.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Cytokine Research, Immunology, Innate Immunity
Primary or Secondary Primary
Gene ID (Entrez) 3570
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.