Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IL6R Antibody (rhPM-1 (Tocilizumab)) - Azide and BSA Free, Novus Biologicals™

Human Monoclonal Antibody
Supplier: Novus Biologicals NBP275192
Description
IL6R Monoclonal specifically detects IL6R in Human samples. It is validated for ELISA, Immunofluorescence, Inhibition Activity.Specifications
IL6R | |
Monoclonal | |
Unconjugated | |
PBS with 0.02% Proclin 300 | |
IL6R | |
This antibody was prepared by CDR grafting of mouse AUK 12-20 antibody onto a human framework regions. | |
0.2 mg | |
3570.0 | |
Human | |
Purified |
ELISA, Immunofluorescence, Inhibition Assays | |
rhPM-1 (Tocilizumab) | |
ELISA 1:100 - 1:2000, Immunofluorescence, Inhibition Activity | |
biosimilar, biosimilars, CD126, CD126 antigen, gp80, IL-6 receptor subunit alpha, IL-6R 1, IL-6R subunit alpha, IL-6R-1, IL6RA, IL-6RA, IL-6R-alpha, interleukin 6 receptor, interleukin 6 receptor alpha subunit, interleukin-6 receptor subunit alpha, Membra | |
Human | |
Protein A purified | |
Primary | |
Tocilizumab binds specifically to both the membrane bound and soluble forms of human IL-6R. The IL6 receptor is a protein complex consisting of an IL-6 receptor subunit (IL6R) and interleukin 6 signal transducer Glycoprotein 130. Interleukin 6 (IL6) is a potent pleiotropic cytokine that regulates cell growth and differentiation and plays an important role in immune response. | |
Store at 4°C for up to 3 months. For longer storage, aliquot and store at -20°C. | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction