Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IL6R Antibody (rhPM-1 (Tocilizumab)) - Azide and BSA Free, Novus Biologicals™

Human Monoclonal Antibody
$268.00 - $729.36
Specifications
Antigen | IL6R |
---|---|
Clone | rhPM-1 (Tocilizumab) |
Dilution | ELISA 1:100 - 1:2000, Immunofluorescence, Inhibition Activity |
Applications | ELISA, Immunofluorescence, Inhibition Assays |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP339823
![]() |
Novus Biologicals
NBP339823 |
0.2 mg |
Each for $729.36
|
|
|||||
NB001495
![]() |
Novus Biologicals
NBP2751920.025MG |
0.025 mg |
Each for $268.00
|
|
|||||
NBP275192
![]() |
Novus Biologicals
NBP275192 |
0.2 mg |
Each for $572.50
|
|
|||||
Description
IL6R Monoclonal specifically detects IL6R in Human samples. It is validated for ELISA, Immunofluorescence, Inhibition Activity.Specifications
IL6R | |
ELISA 1:100 - 1:2000, Immunofluorescence, Inhibition Activity | |
Monoclonal | |
Purified | |
Human | |
This antibody was prepared by CDR grafting of mouse AUK 12-20 antibody onto a human framework regions. | |
Store at 4°C for up to 3 months. For longer storage, aliquot and store at -20°C. |
rhPM-1 (Tocilizumab) | |
ELISA, Immunofluorescence, Inhibition Assays | |
Unconjugated | |
Human | |
IL6R | |
IgG1 κ | |
Tocilizumab binds specifically to both the membrane bound and soluble forms of human IL-6R. The IL6 receptor is a protein complex consisting of an IL-6 receptor subunit (IL6R) and interleukin 6 signal transducer Glycoprotein 130. Interleukin 6 (IL6) is a potent pleiotropic cytokine that regulates cell growth and differentiation and plays an important role in immune response. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title