Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL6R Antibody (rhPM-1 (Tocilizumab)) - Chimeric - Azide and BSA Free, Novus Biologicals™
SDP

Catalog No. NB001496 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.2 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB001496 0.025 mg
NBP275193 0.2 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB001496 Supplier Novus Biologicals Supplier No. NBP2751930.025MG
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

IL6R Monoclonal specifically detects IL6R in Human samples. It is validated for ELISA, Immunofluorescence, Inhibition Activity.

Specifications

Antigen IL6R
Applications ELISA, Immunofluorescence, Inhibition Assays
Classification Monoclonal
Clone rhPM-1 (Tocilizumab)
Conjugate Unconjugated
Dilution ELISA 1:100 - 1:2000, Immunofluorescence, Inhibition Activity
Formulation PBS
Gene Alias biosimilar, biosimilars, CD126, CD126 antigen, gp80, IL-6 receptor subunit alpha, IL-6R 1, IL-6R subunit alpha, IL-6R-1, IL6RA, IL-6RA, IL-6R-alpha, interleukin 6 receptor, interleukin 6 receptor alpha subunit, interleukin-6 receptor subunit alpha, Membrane glycoprotein 80, MGC104991
Gene Symbols IL6R
Host Species Rabbit
Immunogen This antibody was prepared by CDR grafting of mouse AUK 12-20 antibody onto a human framework regions.
Quantity 0.025 mg
Gene ID (Entrez) 3570
Test Specificity Tocilizumab binds specifically to both the membrane bound and soluble forms of human IL-6R. The IL6 receptor is a protein complex consisting of an IL-6 receptor subunit (IL6R) and interleukin 6 signal transducer Glycoprotein 130. Interleukin 6 (IL6) is a potent pleiotropic cytokine that regulates cell growth and differentiation and plays an important role in immune response.
Target Species Human
Content And Storage Store at 4°C for up to 3 months. For longer storage, aliquot and store at -20°C.
Form Purified
Isotype IgG κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.