Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL6R Antibody (rhPM-1 (Tocilizumab)), PerCP, Novus Biologicals™
SDP

Catalog No. NB343039 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB343039 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB343039 Supplier Novus Biologicals Supplier No. NBP275192PCP
Add to Cart
Add to Cart

Human Monoclonal Antibody

IL6R Monoclonal antibody specifically detects IL6R in Human samples. It is validated for ELISA, Immunofluorescence, Inhibition Activity

Specifications

Antigen IL6R
Applications ELISA, Immunofluorescence, Inhibition Assays
Classification Monoclonal
Clone rhPM-1 (Tocilizumab)
Conjugate PerCP
Dilution ELISA, Immunofluorescence, Inhibition Activity
Formulation PBS
Gene Alias CD126, CD126 antigen, gp80, IL-6 receptor subunit alpha, IL-6R 1, IL-6R subunit alpha, IL-6R-1, IL6RA, IL-6RA, IL-6R-alpha, interleukin 6 receptor, interleukin 6 receptor alpha subunit, interleukin-6 receptor subunit alpha, Membrane glycoprotein 80, MGC104991
Host Species Human
Immunogen The original antibody was generated by CDR grafting of mouse PM-1 antibody onto human REI and NEW framework regions to create rhPM-1.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Cytokine Research, Immunology, Innate Immunity
Primary or Secondary Primary
Gene ID (Entrez) 3570
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.