Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL6R Rat anti-Mouse, Clone: D7715A7, Novus Biologicals™
SDP

Catalog No. NB152983 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.2 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB152983 0.2 mg
NB152982 0.025 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB152983 Supplier Novus Biologicals Supplier No. NBP3120100.2MG
Add to Cart
Add to Cart

Rat Monoclonal Antibody

IL6R Monoclonal antibody specifically detects IL6R in Mouse samples. It is validated for Flow Cytometry, Neutralization

Specifications

Antigen IL6R
Applications Flow Cytometry, Neutralization
Classification Monoclonal
Clone D7715A7
Concentration 1 mg/ml
Conjugate Unconjugated
Dilution Flow Cytometry, Block/Neutralize
Formulation PBS
Gene Alias CD126, CD126 antigen, gp80, IL-6 receptor subunit alpha, IL-6R 1, IL-6R subunit alpha, IL-6R-1, IL6RA, IL-6RA, IL-6R-alpha, interleukin 6 receptor, interleukin 6 receptor alpha subunit, interleukin-6 receptor subunit alpha, Membrane glycoprotein 80, MGC104991
Host Species Rat
Immunogen This antibody was raised by immunising Lewis rats with OKT4 cells.
Purification Method Protein A purified
Quantity 0.2 mg
Regulatory Status RUO
Research Discipline Cancer, Cytokine Research, Immunology, Innate Immunity
Primary or Secondary Primary
Gene ID (Entrez) 3570
Target Species Mouse
Content And Storage Store at 4C for up to 3 months. For longer storage, aliquot and store at -20C.
Form Purified
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.