Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IL6R Rat anti-Mouse, Clone: D7715A7, Novus Biologicals™

Rat Monoclonal Antibody
$268.00 - $584.50
Specifications
Antigen | IL6R |
---|---|
Clone | D7715A7 |
Concentration | 1 mg/ml |
Dilution | Flow Cytometry, Block/Neutralize |
Applications | Flow Cytometry, Neutralization |
Description
IL6R Monoclonal antibody specifically detects IL6R in Mouse samples. It is validated for Flow Cytometry, NeutralizationSpecifications
IL6R | |
1 mg/ml | |
Flow Cytometry, Neutralization | |
Unconjugated | |
Rat | |
Cancer, Cytokine Research, Immunology, Innate Immunity | |
PBS | |
3570 | |
IgG2b κ | |
Protein A purified |
D7715A7 | |
Flow Cytometry, Block/Neutralize | |
Monoclonal | |
Purified | |
RUO | |
Mouse | |
CD126, CD126 antigen, gp80, IL-6 receptor subunit alpha, IL-6R 1, IL-6R subunit alpha, IL-6R-1, IL6RA, IL-6RA, IL-6R-alpha, interleukin 6 receptor, interleukin 6 receptor alpha subunit, interleukin-6 receptor subunit alpha, Membrane glycoprotein 80, MGC104991 | |
This antibody was raised by immunising Lewis rats with OKT4 cells. | |
Primary | |
Store at 4C for up to 3 months. For longer storage, aliquot and store at -20C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title