Learn More
Description
Specifications
Specifications
| Antigen | ILDR2 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1.0 ug/ml |
| Formulation | PBS buffer, 2% sucrose |
| Gene Alias | immunoglobulin-like domain containing receptor 2 |
| Host Species | Rabbit |
| Immunogen | The immunogen is a synthetic peptide directed towards the C-terminal region of human ILDR2 (NP_955383.1). Peptide sequence AHLPRLVSRTPGTAPKYDHSYLGSARERQARPEGASRGGSLETPSKRSAQ |
| Purification Method | Affinity purified |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
