Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ILF2 Antibody (OTI6F1), PerCP, Novus Biologicals™
SDP

Catalog No. NB289157 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB289157 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB289157 Supplier Novus Biologicals Supplier No. NBP271036PCP
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

ILF2 Monoclonal antibody specifically detects ILF2 in Human, Mouse, Canine samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence

Specifications

Antigen ILF2
Applications Western Blot, Immunohistochemistry, Immunocytochemistry
Classification Monoclonal
Clone OTI6F1
Conjugate PerCP
Dilution Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence
Formulation PBS
Gene Alias interleukin enhancer binding factor 2, 45kD, interleukin enhancer binding factor 2, 45kDa, interleukin enhancer-binding factor 2, MGC8391, NF45PRO3063, Nuclear factor of activated T-cells 45 kDa, nuclear factor of activated T-cells, 45-kDa
Host Species Mouse
Immunogen Full-length protein expressed in 293T cell transfected with human ILF2 expression vector
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3608
Target Species Human, Mouse, Canine
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.