Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ILF2 Antibody, PerCP, Novus Biologicals™
SDP

Catalog No. NB291020 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB291020 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB291020 Supplier Novus Biologicals Supplier No. NBP171902PCP
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ILF2 Polyclonal antibody specifically detects ILF2 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin)

Specifications

Antigen ILF2
Applications Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate PerCP
Dilution Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin
Formulation PBS
Gene Alias interleukin enhancer binding factor 2, 45kD, interleukin enhancer binding factor 2, 45kDa, interleukin enhancer-binding factor 2, MGC8391, NF45PRO3063, Nuclear factor of activated T-cells 45 kDa, nuclear factor of activated T-cells, 45-kDa
Host Species Rabbit
Immunogen The immunogen for this product maps to a region between residue 340 and 390 of human Nuclear Factor of Activated T-cells 45 kDa using the numbering given in entry NP_004506.2 (GeneID 3608).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3608
Target Species Human, Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.