Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ILK Antibody (OTI2H3), Alexa Fluor™ 594, Novus Biologicals™
SDP

Catalog No. NP271037A94 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NP271037A94 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NP271037A94 Supplier Novus Biologicals Supplier No. NBP271037AF594
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

ILK Monoclonal specifically detects ILK in Human, Mouse, Rat samples. It is validated for Western Blot.

Specifications

Antigen ILK
Applications Western Blot
Classification Monoclonal
Clone OTI2H3
Conjugate Alexa Fluor 594
Dilution Western Blot
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias 59 kDa serine/threonine-protein kinase, DKFZp686F1765, EC 2.7.11.1, ILK1, ILK-1, ILK2, ILK-2, integrin-linked kinase, integrin-linked kinase-2, integrin-linked protein kinase, P59, p59ILK
Gene Symbols ILK
Host Species Mouse
Immunogen Recombinant protein fragment corresponding to amino acids 1-244 of human ILK (NP_004508) produced in E.coli.
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 3611
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.