Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ILK Antibody (OTI2H3) - Azide and BSA Free, Novus Biologicals™
SDP

Catalog No. NBP271037 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP271037 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP271037 Supplier Novus Biologicals Supplier No. NBP271037
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

ILK Monoclonal specifically detects ILK in Human, Mouse, Rat samples. It is validated for Western Blot.

Specifications

Antigen ILK
Applications Western Blot
Classification Monoclonal
Clone OTI2H3
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1:2000
Formulation Lyophilized from PBS (pH 7.3) with 8% Trehalose with No Preservative
Gene Alias 59 kDa serine/threonine-protein kinase, DKFZp686F1765, EC 2.7.11.1, ILK1, ILK-1, ILK2, ILK-2, integrin-linked kinase, integrin-linked kinase-2, integrin-linked protein kinase, P59, p59ILK
Gene Symbols ILK
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 1-244 of human ILK (NP_004508) produced in E.coli.
Molecular Weight of Antigen 51.2 kDa
Purification Method Affinity Purified
Quantity 100 μg
Research Discipline Phospho Specific, Protein Kinase, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 3611
Reconstitution Reconstitute with PBS (pH 7.3). To use this carrier-free antibody for conjugation experiments, another round of desalting is highly recommended.
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Lyophilized
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.