Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ILK Rabbit anti-Human, Mouse, Rat, Clone: 5R10U3, Novus Biologicals™
SDP

Catalog No. NB167970 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB167970 20 μg
NB167969 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB167970 Supplier Novus Biologicals Supplier No. NBP31643920UL
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

ILK Monoclonal antibody specifically detects ILK in Human, Mouse, Rat samples. It is validated for Western Blot

Specifications

Antigen ILK
Applications Western Blot
Classification Monoclonal
Clone 5R10U3
Conjugate Unconjugated
Formulation PBS, 0.05% BSA, 50% glycerol, pH7.3
Gene Alias 59 kDa serine/threonine-protein kinase, DKFZp686F1765, EC 2.7.11.1, ILK1, ILK-1, ILK2, ILK-2, integrin-linked kinase, integrin-linked kinase-2, integrin-linked protein kinase, P59, p59ILK
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 350-452 of human ILK (Q13418).
Purification Method Affinity purified
Quantity 20 μg
Regulatory Status RUO
Research Discipline Protein Kinase, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 3611
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.