Learn More
Description
- Monomeric and bacterial
- Molecular weight: 54.5kd
- Expressed in E.Coli
- Inactivated by heating at 80°C for 15 minutes
- Active in PCR buffers
- Activity: 10U/μL
- Free of contaminating endonucleases, double-stranded exonucleases and ribonucleases
- >95% pure by SDS-PAGE
Specifications
Specifications
| Purity | >95% by SDS-PAGE |
| Source | Bacteria |
| Quantity | 5000 U |
| Product Type | Exonuclease I |
| Molecular Weight (g/mol) | 54.5kd |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.