Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva ExoProStar DNA and Sequence Reaction Clean-up

Catalog No. p-4390055 Shop All Cytiva Products
Click to view available options
No. of Reactions:
20
100
500
2000
5000

Optimized to purify DNA and sequencing set up reactions quickly, efficiently and reliably. illustra™ ExoProStar DNA and Sequence Reaction Clean-up combines enzymatic PCR clean up technology with illustra Exonuclease I and Alkaline Phosphatase to remove unincorporated primers and dNTPs.

Enzymes optimized to work together for high efficiency removal of unincorporated primers and dNTPs. Enzymes provided in two separate tubes, just two simple pipetting steps are needed to prepare the reaction.

  • Fast 30-minute protocol
  • Scalable for different reaction sizes
  • No loss of PCR product
  • Easy to automate
  • Complete heat inactivation of the enzymes within 15 minutes

TRUSTED_SUSTAINABILITY

Specifications

Description Enzymatic PCR clean up technology with illustra Exonuclease I and Alkaline Phosphatase to remove unincorporated primers and dNTPs. For maximum flexibility comes in two separate tubes.
For Use With (Application) Optimized to purify DNA and sequencing set up reactions quickly, efficiently and reliably
Includes One tube of Alkaline Phosphatase and Exonuclease I
No. of Reactions 20
Product Type illustra ExoProStar
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.