Learn More
Description
- puReTaq DNA polymerase and high-purity reagents provide robust performance with very low levels of contaminating DNA
- Manufacturing process locks in performance for excellent reproducibility with less risk of pipetting errors and contamination
- Long-term ambient-temperature stability simplifies shipping and removes any need for freezer storage
- Verified for real-time PCR
- Ready-To-Go means simply add template DNA and primers and cycle
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
| Content And Storage | Room temperature stable beads containing ∼ 2.5 units of recombinant PuReTaq DNA Polymerase, dATP, dCTP, dGTP, dTTP, stabilizers, BSA and reaction Buffer, When a bead is reconstituted to a 25μL final volume, the concentration of each dNTP |
| Format | 0.2mL Tubes/plate |
| Quantity | 1 Kit |
| For Use With (Application) | Standard PCR |
| No. of Reactions | 96 |
| Volume | 200μL |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
