Learn More
Description
Includes
10x reaction buffer: 200mM Tris-HCl, pH 8.0, 100mM MgCl2; Dilution buffer: 50mM Tris-HCl, pH 8.0
- Bacterial
- Expressed in E.Coli
- Inactivated by heating at 75°C for 5 minutes
- Active in PCR buffers and restriction endonuclease buffers
- Activity: 1U/μL
- Purified to homogeneity
- Free of all contaminating endonucleases, exonucleases and ribonucleases
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
| Purity | Purified to homogeneity |
| Source | Bacteria |
| Quantity | 5000 U |
| Product Type | Alkaline Phosphatase |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
