Learn More
Cytiva Solution dNTP Sets

Use dNTP sets for optimum performance and consistency. Cytiva Lifescience™ illustra™ Solution dNTP Sets are free from DNase, RNase, and nicking enzyme activity.
$401.50 - $3598.10
Specifications
Format | Liquid |
---|---|
pH | 7 |
Product Type | dNTP set |
Concentration | 100 mM |
Purity | >99% |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
45-001-454
![]() |
Cytiva
28406551 |
4 x 25 μm |
Each for $401.50
|
|
|||||
45-001-455
![]() |
Cytiva
28406552 |
4 x 100 μm |
Each for $1,323.30
|
|
|||||
45-001-456
|
Cytiva
28406553 |
4 x 500 μm |
Each for $3,598.10
|
|
|||||
Description
- Greater than 99% triphosphate purity (by HPLC) for optimum performance and consistency
- Buffer-free and ready-to-use solutions in multiple formats
- Functionally tested for long PCR and sequencing
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Order Info
Custom formulation, testing, packaging, and labeling are available
Specifications
Liquid | |
dNTP set | |
>99% | |
Functionally tested for long PCR and sequencing, Nucleic Acid amplification, labelling, mutagenesis, cDNA synthesis and expression profiling |
7 | |
100 mM | |
Colorless |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.