Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ILT1/CD85h/LILRA2 Antibody (600007), HRP, Novus Biologicals™
SDP

Catalog No. NB347024 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB347024 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB347024 Supplier Novus Biologicals Supplier No. FAB6364H
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

ILT1/CD85h/LILRA2 Monoclonal specifically detects ILT1/CD85h/LILRA2 in Human samples. It is validated for CyTOF, Flow Cytometry.

Specifications

Antigen ILT1/CD85h/LILRA2
Applications CyTOF, Flow Cytometry
Classification Monoclonal
Clone 600007
Conjugate HRP
Dilution Flow Cytometry, CyTOF-ready
Formulation PBS
Gene Alias CD85 antigen-like family member H, CD85h, CD85h antigen, ILT-1, ILT1CD85H, Leukocyte immunoglobulin-like receptor 7, leukocyte immunoglobulin-like receptor subfamily A member 2, leukocyte immunoglobulin-like receptor subfamily A member 2 soluble, leukocyte immunoglobulin-like receptor, subfamily A (with TM domain), member 2, LIR7LIR-7Immunoglobulin-like transcript 1
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human LILRA2/CD85h/ILT1, Pro17-Ser437, Accession # Q8N149
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11027
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.