Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ILT11/LILRA5 Antibody (711828), Alexa Fluor™ 532, Novus Biologicals™
SDP

Catalog No. NB375823 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB375823 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB375823 Supplier Novus Biologicals Supplier No. FAB6754X
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

ILT11/LILRA5 Monoclonal antibody specifically detects ILT11/LILRA5 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, CyTOF

Specifications

Antigen ILT11/LILRA5
Applications Flow Cytometry, Immunocytochemistry, CyTOF
Classification Monoclonal
Clone 711828
Conjugate Alexa Fluor 532
Dilution Flow Cytometry, Immunocytochemistry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias CD85, CD85 antigen-like family member F, CD85F, CD85f antigen, ILT-11, ILT11CD85f, Immunoglobulin-like transcript 11, leukocyte Ig-like receptor 9, Leukocyte immunoglobulin-like receptor 9, leukocyte immunoglobulin-like receptor subfamily A member 5, leukocyte immunoglobulin-like receptor subfamily A member 5 soluble, leukocyte immunoglobulin-like receptor, subfamily A (with TM domain), member 5, leukocyte immunoglobulin-like receptor, subfamily B (with TM and ITIM domains), LILRB7, LIR-9, LIR9immunoglobulin-like transcript 11 protein, member 7
Host Species Mouse
Immunogen Chinese hamster ovary cell line CHO-derived recombinant human LILRA5/CD85f, Gly42-Arg268, Accession # A6NI73
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 353514
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.