Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ILT2/CD85j/LILRB1 Antibody (292305), mFluor Violet 610 SE, Novus Biologicals™
SDP

Catalog No. NB349382 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB349382 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB349382 Supplier Novus Biologicals Supplier No. FAB20171MFV610
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

ILT2/CD85j/LILRB1 Monoclonal specifically detects ILT2/CD85j/LILRB1 in Human samples. It is validated for CyTOF, Flow Cytometry, Western Blot.

Specifications

Antigen ILT2/CD85j/LILRB1
Applications CyTOF, Flow Cytometry, Western Blot
Classification Monoclonal
Clone 292305
Conjugate mFluor Violet 610 SE
Dilution Western Blot, Flow Cytometry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias CD85 antigen-like family member J, CD85j, CD85j antigen, Ig-like transcript 2, ILT-2, ILT2FLJ37515, Immunoglobulin-like transcript 2, leukocyte Ig-like receptor-1, Leukocyte immunoglobulin-like receptor 1, leukocyte immunoglobulin-like receptor subfamily B member 1, leukocyte immunoglobulin-like receptor subfamily B member 1 soluble isoform, leukocyte immunoglobulin-like receptor, subfamily B (with TM and ITIM domains), LIR1LIR-1MIR-7CD85MIR7CD85 antigen, member 1, Monocyte/macrophage immunoglobulin-like receptor 7
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human LILRB1/CD85j/ILT2, Gly24-His458, Accession # Q8NHL6
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Gene ID (Entrez) 10859
Target Species Human
Content And Storage Store at 4C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.