Learn More
Description
Specifications
Specifications
| Antigen | ILT3/CD85k/LILRB4 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1.0 ug/ml |
| Formulation | PBS buffer, 2% sucrose |
| Gene Alias | CD85 antigen-like family member K, CD85k, CD85k antigen, ILT-3, ILT3CD85K, leukocyte immunoglobulin-like receptor, subfamily B (with TM and ITIM domains), LIR5LILRB5, LIR-5subfamily B, member 4, member 4 |
| Host Species | Rabbit |
| Immunogen | The immunogen for Anti-ILT3/CD85k/LILRB4 antibody is: synthetic peptide directed towards the C-terminal region of Human LIRB4. Peptide sequence RQGKHRTLAQRQADFQRPPGAAEPEPKDGGLQRRSSPAADVQGENFCAAV |
| Purification Method | Affinity purified |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
