Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ILT4/CD85d/LILRB2 Antibody (287219R), PerCP, Novus Biologicals™
SDP

Catalog No. NB366527 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB366527 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB366527 Supplier Novus Biologicals Supplier No. FAB2078RC
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

ILT4/CD85d/LILRB2 Monoclonal specifically detects ILT4/CD85d/LILRB2 in Human samples. It is validated for CyTOF, Flow Cytometry, Neutralization, Western Blot.

Specifications

Antigen ILT4/CD85d/LILRB2
Applications CyTOF, Flow Cytometry, Neutralization, Western Blot
Classification Monoclonal
Clone 287219R
Conjugate PerCP
Dilution Western Blot, Flow Cytometry, Neutralization, CyTOF-ready
Formulation PBS
Gene Alias CD85 antigen-like family member D, CD85d antigen, Ig-like transcript 4, ILT-4, ILT4CD85d, Immunoglobulin-like transcript 4, Leukocyte immunoglobulin-like receptor 2, leukocyte immunoglobulin-like receptor subfamily B member 2, leukocyte immunoglobulin-like receptor subfamily B member 2 soluble isoform, leukocyte immunoglobulin-like receptor, subfamily B (with TM and ITIM domains), LIR2CD85D, LIR-2subfamily A (with TM domain), member 6, member 2, MIR10LILRA6, Monocyte/macrophage immunoglobulin-like receptor 10
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human LILRB2/CD85d/ILT4, Gly24-His458, Accession # ACT64556
Purification Method Protein A or G purified from cell culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Gene ID (Entrez) 10288
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.