Learn More
Description
Specifications
Specifications
| Antigen | ILT5/CD85a/LILRB3 |
| Applications | ELISA |
| Classification | Monoclonal |
| Clone | 123 |
| Conjugate | DyLight 350 |
| Dilution | ELISA |
| Formulation | 50mM Sodium Borate |
| Gene Alias | CD85 antigen-like family member A, CD85a antigen, HL9Immunoglobulin-like transcript 5, ILT-5, ILT5CD85a, Leukocyte immunoglobulin-like receptor 3, leukocyte immunoglobulin-like receptor subfamily B member 3, leukocyte immunoglobulin-like receptor, subfamily B (with TM and ITIM domains), LIR3CD85A, LIR-3MGC138403, member 3, Monocyte inhibitory receptor HL9, PIRB |
| Host Species | Rabbit |
| Immunogen | This antibody was obtained from a rabbit immunized with purified, recombinant Human ILT5/CD85a/LILRB3 (NP_006855.2; Met1-Glu443). |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
