Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ILT5/CD85a/LILRB3 Antibody (4F9), PE, Novus Biologicals™
SDP

Catalog No. NB164329PE Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB164329PE 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB164329PE Supplier Novus Biologicals Supplier No. NB10064329PE
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

ILT5/CD85a/LILRB3 Monoclonal specifically detects ILT5/CD85a/LILRB3 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen).

Specifications

Antigen ILT5/CD85a/LILRB3
Applications Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen)
Classification Monoclonal
Clone 4F9
Conjugate PE
Dilution Flow Cytometry
Formulation PBS with 0.05% Sodium Azide
Gene Alias CD85 antigen-like family member A, CD85a antigen, HL9Immunoglobulin-like transcript 5, ILT-5, ILT5CD85a, Leukocyte immunoglobulin-like receptor 3, leukocyte immunoglobulin-like receptor subfamily B member 3, leukocyte immunoglobulin-like receptor, subfamily B (with TM and ITIM domains), LIR3CD85A, LIR-3MGC138403, member 3, Monocyte inhibitory receptor HL9, PIRB
Gene Symbols LILRB3
Host Species Mouse
Immunogen Monocyte derived dendritic cells
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11025
Test Specificity NB100-64329 is specific for human CD85a (leucocyte immunoglobulin-like receptor, subfamily B, member 3/ LILRB3), also known as ILT-5, expressed by monocytes, dendritic cells, macrophages, subpopulation of T lymphocytes, B cells and at high levels on plasma cells. CD85a is a 631 amino acid type I transmembrane glycoprotein, which contains four immunoreceptor tyrosine-based inhibition motif (ITIM) sequences within a long cytoplasmic tail. Phosphorylation of the tyrosine residues within ITIMs is known to enable the binding and activation of protein tyrosine phosphatases, which act as cell signalling modulators and inhibitors of cell activation.
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.