Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ILT6/CD85e/LILRA3 Antibody, mFluor Violet 610 SE, Novus Biologicals™
SDP

Catalog No. NB322304 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB322304 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB322304 Supplier Novus Biologicals Supplier No. NBP300150MFV610
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ILT6/CD85e/LILRA3 Polyclonal antibody specifically detects ILT6/CD85e/LILRA3 in Human samples. It is validated for ELISA

Specifications

Antigen ILT6/CD85e/LILRA3
Applications ELISA
Classification Polyclonal
Conjugate mFluor Violet 610 SE
Dilution ELISA
Formulation 50mM Sodium Borate
Gene Alias CD85 antigen-like family member E, CD85e antigen, HM31, ILT-6, ILT6CD85e, Immunoglobulin-like transcript 6, leucocyte immunoglobulin-like receptor, Leukocyte immunoglobulin-like receptor 4, leukocyte immunoglobulin-like receptor A3, leukocyte immunoglobulin-like receptor subfamily A member 3, leukocyte immunoglobulin-like receptor, subfamily A (without TM domain), member3, LIR4CD85E, LIR-4HM43e3, Monocyte inhibitory receptor HM43/HM31
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant Human ILT6/CD85e/LILRA3 (Accession#: AAH28208.1; Met1-Glu439)
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11026
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.