Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ILT6/CD85e/LILRA3 Rabbit anti-Human, Clone: 60, Novus Biologicals™
SDP

Catalog No. NB021919 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB021919 100 μg
NB021920 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB021919 Supplier Novus Biologicals Supplier No. NBP290228100UL
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

ILT6/CD85e/LILRA3 Monoclonal specifically detects ILT6/CD85e/LILRA3 in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen ILT6/CD85e/LILRA3
Applications Flow Cytometry
Classification Monoclonal
Clone 60
Conjugate Unconjugated
Dilution Flow Cytometry 1:25-1:100
Formulation 0.2 um filtered solution in PBS
Gene Alias CD85 antigen-like family member E, CD85e antigen, HM31, ILT-6, ILT6CD85e, Immunoglobulin-like transcript 6, leucocyte immunoglobulin-like receptor, Leukocyte immunoglobulin-like receptor 4, leukocyte immunoglobulin-like receptor A3, leukocyte immunoglobulin-like receptor subfamily A member 3, leukocyte immunoglobulin-like receptor, subfamily A (without TM domain), member3, LIR4CD85E, LIR-4HM43e3, Monocyte inhibitory receptor HM43/HM31
Host Species Rabbit
Immunogen Recombinant Human ILT6/CD85e/LILRA3 Protein
Purification Method Protein A purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11026
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.