Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ILT7/CD85g/LILRA4 Antibody (656688) [DyLight 755], Novus Biologicals™
SDP

Catalog No. NB376755 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB376755 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB376755 Supplier Novus Biologicals Supplier No. FAB6287Z
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

ILT7/CD85g/LILRA4 Monoclonal antibody specifically detects ILT7/CD85g/LILRA4 in Human samples. It is validated for Flow Cytometry, CyTOF

Specifications

Antigen ILT7/CD85g/LILRA4
Applications Flow Cytometry, CyTOF
Classification Monoclonal
Clone 656688
Conjugate DyLight 755
Dilution Flow Cytometry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias CD85 antigen-like family member G, CD85g, CD85g antigen, ILT-7, ILT7MGC129598, Immunoglobulin-like transcript 7, leukocyte immunoglobulin-like receptor subfamily A member 4, leukocyte immunoglobulin-like receptor, subfamily A (with TM domain), member 4, leukocyte immunoglobulin-like receptor, subfamily A (without TM domain), member4, MGC129597
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human LILRA4/CD85g/ILT7, Glu24-Asn446, Accession # P59901
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23547
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.