Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ILT8/CD85b/LILRA6 Antibody (921330), PE/Cy5.5, Novus Biologicals™
SDP

Catalog No. NB310673 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB310673 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB310673 Supplier Novus Biologicals Supplier No. FAB8656PECY55
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

ILT8/CD85b/LILRA6 Monoclonal antibody specifically detects ILT8/CD85b/LILRA6 in Human samples. It is validated for Flow Cytometry

Specifications

Antigen ILT8/CD85b/LILRA6
Applications Flow Cytometry
Classification Monoclonal
Clone 921330
Conjugate PE-Cyanine5.5
Dilution Flow Cytometry
Formulation PBS
Gene Alias CD85b;ILT5;ILT8;ILT-8;LILRB3;LILRB6
Host Species Mouse
Immunogen HEK293 human embryonic kidney cell line-derived recombinant human LILRA6/CD85b, Met1-Asn447, Accession # Q6PI73
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79168
Target Species Human
Content And Storage Store at 4C in the dark. Do not freeze.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.