Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ILVBL Antibody, Alexa Fluor™ 488, Novus Biologicals™
SDP

Catalog No. NBP178433X Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP178433X 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP178433X Supplier Novus Biologicals Supplier No. NBP178433AF488
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ILVBL Polyclonal specifically detects ILVBL in Human samples. It is validated for Western Blot.

Specifications

Antigen ILVBL
Applications Western Blot
Classification Polyclonal
Conjugate Alexa Fluor 488
Dilution Western Blot
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Accession No. A1L0T0
Gene Alias acetolactate synthase homolog, acetolactate synthase-like protein, AHASFLJ39061,209L8, EC 2.2.1, EC 2.2.1.-, ILV2H, ilvB (bacterial acetolactate synthase)-like, IlvB-like protein, MGC1269, MGC19535
Gene Symbols ILVBL
Host Species Rabbit
Immunogen A synthetic peptide made to an internal portion of the human ILVBL protein (between residues 350-500) [UniProt A1L0T0]
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Epigenetics
Primary or Secondary Primary
Gene ID (Entrez) 10994
Target Species Human
Content And Storage Store at 4C in the dark.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.