Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ILVBL Antibody (OTI8B12), Alexa Fluor™ 405, Novus Biologicals™
SDP

Catalog No. NB27161A405 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB27161A405 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB27161A405 Supplier Novus Biologicals Supplier No. NBP271661AF405

Mouse Monoclonal Antibody

ILVBL Monoclonal specifically detects ILVBL in Human, Mouse, Rat, Canine, Primate, Monkey samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen ILVBL
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone OTI8B12
Conjugate Alexa Fluor 405
Dilution Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias acetolactate synthase homolog, acetolactate synthase-like protein, AHASFLJ39061,209L8, EC 2.2.1, EC 2.2.1.-, ILV2H, ilvB (bacterial acetolactate synthase)-like, IlvB-like protein, MGC1269, MGC19535
Gene Symbols ILVBL
Host Species Mouse
Immunogen Full length recombinant protein of human ILVBL (NP_006835) produced in HEK293T cells.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Epigenetics
Primary or Secondary Primary
Gene ID (Entrez) 10994.0
Target Species Human, Mouse, Rat, Canine, Primate, Monkey
Content And Storage Store at 4°C in the dark.
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.