Learn More
Description
- Uncharged IMAC resin allows possibility to charge with metal ion of choice for optimized selectivity
- Used for ions such as histidine-tagged proteins but also untagged recombinant or native proteins
- High protein binding capacity
- Coupling chemistry and novel ligand designed to give minimal ligand leakage
- Available HiTrap™ and HiPrep™ pre-packed formats for easy scale-up and process development
Specifications
Specifications
| Quantity | 25 mL |
| For Use With (Application) | For Purifying Proteins and Peptides That Have an Affinity for Metal Ions |
| Product Type | IMAC Sepharose 6 Fast Flow Media |
| Particle Size | 90 μm |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.