Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Imatinib mesylate, 98%

Catalog No. p-7077869
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

CAS: 220127-57-1 | C30H35N7O4S | 589.72 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 220127-57-1
Molecular Formula C30H35N7O4S
Molecular Weight (g/mol) 589.72
MDL Number MFCD04307699
InChI Key YLMAHDNUQAMNNX-UHFFFAOYSA-N
Synonym imatinib mesylate, gleevec, glivec, imatinib mesilate, imatinib methanesulfonate, imatinib mesylate sti571, imatinib monomesylate, unii-8a1o1m485b, imatinib, methanesulfonate salt
PubChem CID 123596
ChEBI CHEBI:31690
IUPAC Name methanesulfonic acid;4-[(4-methylpiperazin-1-yl)methyl]-N-[4-methyl-3-[(4-pyridin-3-ylpyrimidin-2-yl)amino]phenyl]benzamide
SMILES CS(O)(=O)=O.CN1CCN(CC2=CC=C(C=C2)C(=O)NC2=CC=C(C)C(NC3=NC=CC(=N3)C3=CC=CN=C3)=C2)CC1

Specifications

Melting Point 209.0°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass Bottle
Quantity 1 g
Solubility Information Solubility in water: soluble.
Formula Weight 589.71
Percent Purity 98%
Chemical Name or Material Imatinib mesylate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.