Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Imidazole, ULTROL™ Grade, Calbiochem™,

Catalog No. p-4953790 Shop All MilliporeSigma Products
Click to view available options
Quantity:
25 g
100 g

Useful in the pH range of 6.2 to 7.8

TRUSTED_SUSTAINABILITY

Specifications

CAS 288-32-4
Quantity 25 g
Molecular Formula C3H4N2
MDL Number MFCD00005183
Synonym 1,3-Diaza-2,4-cyclopentadiene
InChI Key RAXXELZNTBOGNW-UHFFFAOYSA-N
SMILES N1C=CN=C1
IUPAC Name 1H-imidazole
Molecular Weight (g/mol) 68.079
Grade ULTROL™ Grade
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.