Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Imiloxan hydrochloride
GREENER_CHOICE

Catalog No. 098610
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
09-861-0 10 mg
09-865-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 09-861-0 Supplier Tocris Bioscience™ Supplier No. 0986/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Highly selective α2B antagonist

A moderately potent, but highly selective α2-adrenoceptor antagonist, the only reported selective antagonist at the α2B-adrenoceptor not having potent α1-adrenoceptor antagonist activity.

Chemical Identifiers

CAS 81167-22-8
Molecular Formula C14H17ClN2O2

Specifications

Quantity 10 mg
Formula Weight 280.75
Purity >98%
Molecular Weight (g/mol) 280.752
Chemical Name or Material Imiloxan hydrochloride
Recommended Storage Desiccate at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.