Learn More
Cytiva Immobiline™ DryStrip Kit and Components
Catalog No. 45000435 Shop All Cytiva Products
Change view
Click to view available options
Type:
Anode
Cathode
Dry Strip Slinger
Immobiline DryStrip Kit
Sample Cup Bar
Tray and Electrode Holder
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type
45-000-435 Anode
45-001-798 Immobiline DryStrip Kit
45-001-799 Tray and Electrode Holder
45-002-003 Sample Cup Bar
45-001-801 Dry Strip Slinger
45-000-436 Cathode
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 45-000-435 Supplier Cytiva Supplier No. 18101866
Precast Immobiline DryStrip gels for running the first dimension of a 2-D electrophoresis experiment with the Multiphor II electrophoresis system
Run up to 12 Immobiline DryStrip strips simultaneously with up to 100μL on each Immobiline DryStrip.
  • The instruction manual presents all necessary information for the entire method for horizontal 2-D electrophoresis. By appropriate adaptations, the SDS electrophoresis can also be performed using vertical electrophoresis equipment
  • Immobiline DryStrip kit facilitates sample application and running and equilibration of Immobiline DryStrip for the first dimension in 2-D (two-dimensional) electrophoresis
  • Isoelectric focusing with an immobilized pH gradient (IPG), makes true isoelectric focusing possible and significantly improves the reproducibility of the spot distribution along the pH gradient axis of 2-D maps
  • Individual components are also available separately
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Multiphor II Electrophoresis System
Type Anode