Learn More
Invitrogen™ IMMP2L Polyclonal Antibody

Description
This antibody is predicted to react with mouse based on sequence homology.
Specifications
Specifications
| Antigen | IMMP2L |
| Applications | Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.09% sodium azide; pH 7.4 |
| Gene | IMMP2L |
| Gene Accession No. | Q96T52 |
| Gene Alias | AI853880; IMMP2L; IMMP2L-IT1; IMP2; IMP2 inner mitochondrial membrane peptidase-like; IMP2 inner mitochondrial membrane peptidase-like (S. cerevisiae); IMP2-LIKE; IMP2-like protein; inner mitochondrial membrane peptidase 2 like; inner mitochondrial membrane peptidase subunit 2; mitochondrial inner membrane protease subunit 2 |
| Gene Symbols | IMMP2L |
| Host Species | Rabbit |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.