Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Immuno Tubes and Stoppers

Catalog No. 12565146
Click to view available options
Volume (Metric):
12 mL
5 mL
Not Applicable
Material:
Polyethylene
Polystyrene
Diameter (Metric) Outer:
12 mm
15 mm
Product Type:
Immuno Tubes and Stopper
Stopper
This item is not returnable. View return policy
This item is not returnable. View return policy

Reproduce results time after time with these immuno tubes and stoppers, ideal for ELISA, solid phase RIA and liquid phase RIA assays.

Reproduce results time after time with Thermo Scientific™ Immuno Tubes and Stoppers, ideal for ELISA, solid phase RIA and liquid phase RIA assays. These products are backed by industry-leading quality standards and certifications.

Features:

  • Offered in choice of three surfaces for optimal binding
  • Three tube sizes with stoppers available (ordered separately)
  • Fits standard equipment

Choice of Surface:

  • Thermo Scientific™ PolySorp™: a hydrophobic surface that preferentially binds biomolecules that have hydrophobic domains
  • Thermo Scientific™ MaxiSorp™: a hydrophilic surface that is optimized for binding IgG and also displays increased binding of many other proteins and biomolecules that possess hydrophilic/hydrophobic character
  • Thermo Scientific™ Mini-Sorp™: a polyethylene-based material that provides a low-binding surface

Liquid Phase Immunoassay Tubes (Cat. Nos. 443990, 468608):

  • MiniSorp surface
  • Molded from special polyethylene formulation
  • For liquid phase immunoasssays, including RIA, for storing of reagents or collecting chromatographic fractions

Solid Phase Immunoassay Tubes:

  • PolySorp and MaxiSorp surfaces available for choice of optimal binding
  • For solid phase immuno techniques such as IRMA, ELISA and ILMA
  • Certified binding homogeneity
  • High-quality polystyrene for optimal quality
  • StarTube design gives higher sensitivity and faster assay times
  • Enhances assay sensitivity and reduces incubation times

Recommended for:

  • ELISA
  • Solid Phase RIA
  • Liquid Phase RIA
  • Sample storage
TRUSTED_SUSTAINABILITY

Specifications

Product Type Immuno Tubes and Stopper
Material Polystyrene
Surface MaxiSorp
Length (Metric) 75 mm
Diameter (Metric) Outer 12 mm
No. per Pack 100
No. per Case 3000
Volume (Metric) 5 mL
For Use With (Equipment) Immuno Tube
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.