Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Thermo Scientific™ Immuno Tubes and Stoppers
Click to view available options
Volume (Metric):
12 mL
5 mL
Not Applicable
Material:
Polyethylene
Polystyrene
Diameter (Metric) Outer:
12 mm
15 mm
Product Type:
Immuno Tubes and Stopper
Stopper
Description
Reproduce results time after time with Thermo Scientific™ Immuno Tubes and Stoppers, ideal for ELISA, solid phase RIA and liquid phase RIA assays. These products are backed by industry-leading quality standards and certifications.
Features:
- Offered in choice of three surfaces for optimal binding
- Three tube sizes with stoppers available (ordered separately)
- Fits standard equipment
Choice of Surface:
- Thermo Scientific™ PolySorp™: a hydrophobic surface that preferentially binds biomolecules that have hydrophobic domains
- Thermo Scientific™ MaxiSorp™: a hydrophilic surface that is optimized for binding IgG and also displays increased binding of many other proteins and biomolecules that possess hydrophilic/hydrophobic character
- Thermo Scientific™ Mini-Sorp™: a polyethylene-based material that provides a low-binding surface
Liquid Phase Immunoassay Tubes (Cat. Nos. 443990, 468608):
- MiniSorp surface
- Molded from special polyethylene formulation
- For liquid phase immunoasssays, including RIA, for storing of reagents or collecting chromatographic fractions
Solid Phase Immunoassay Tubes:
- PolySorp and MaxiSorp surfaces available for choice of optimal binding
- For solid phase immuno techniques such as IRMA, ELISA and ILMA
- Certified binding homogeneity
- High-quality polystyrene for optimal quality
- StarTube design gives higher sensitivity and faster assay times
- Enhances assay sensitivity and reduces incubation times
Recommended for:
- ELISA
- Solid Phase RIA
- Liquid Phase RIA
- Sample storage

Specifications
Specifications
Product Type | Stopper |
Material | Polyethylene |
Diameter (Metric) Outer | 15 mm |
No. per Pack | 300 |
No. per Case | 2400 |
Volume (Metric) | Not Applicable |
For Use With (Equipment) | 15 mm Nunc-Immuno Tubes |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction