Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IMP Dehydrogenase 2/IMPDH2 Antibody, Novus Biologicals™
SDP

Catalog No. NBP227195SS Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP227195SS 0.025 mg
NBP227195 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP227195SS Supplier Novus Biologicals Supplier No. NBP227195SS
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

IMP Dehydrogenase 2/IMPDH2 Polyclonal specifically detects IMP Dehydrogenase 2/IMPDH2 in Human, Mouse, Rat, Porcine, Amphibian, Bovine, Canine, Chicken, Chinese Hamster, Equine, Guinea Pig, Opossum, Primate, Rabbit samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen IMP Dehydrogenase 2/IMPDH2
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Western Blot 2-4ug/ml∼, Immunohistochemistry 5ug/ml, Immunohistochemistry-Paraffin
Formulation 0.2 ml PBS and 0.05% BSA with 0.05% Sodium Azide
Gene Accession No. P12268
Gene Alias EC 1.1.1.205, IMP (inosine 5'-monophosphate) dehydrogenase 2, IMP (inosine monophosphate) dehydrogenase 2, IMP dehydrogenase 2, IMPD 2, IMPD2IMP oxireductase 2, IMPDH 2, IMPDH-II, inosine 5' phosphate dehydrogenase 2, inosine monophosphate dehydrogenase type II, inosine-5'-monophosphate dehydrogenase 2
Gene Symbols IMPDH2
Host Species Rabbit
Immunogen Amino acids 221-248 from human IMPDH was used as the immunogen for this antibody.
Purification Method Protein A purified
Quantity 0.025 mg
Regulatory Status RUO
Research Discipline Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 3615
Test Specificity The amino acid sequence used as immunogen for this antibody is 100% homologous in human, mouse, rat, sumatran orangutan, white-cheeked gibbon, rhesus monkey, new world monkey, panda, cow, guinea pig, dog, Chinese hamster, zebrafish, horse, chicken, african bush elephant, wild turkey, opossum, platypus, cichlid fish, rabbit, chimpanzee, salmon, pig, zebra finch, and frog.This antibody will detect both IMPDH1 and IMPDH2 in test samples. Additionally, there are multiple isoforms of the IMPDH protein. Researchers are encouraged to use a tool such as NCBI BLAST to determine theoretical sizes of, and cross reaction with, the various isoforms.
Target Species Human, Mouse, Rat, Pig, Amphibian, Bovine, Canine, Chicken, Chinese Hamster, Equine, Guinea Pig, Opossum, Other
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.